Learn More
Sino Biological Cynomolgus NT5E PAb 100ul (Sino Biological)
Supplier:  Sino Biological 90192RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus CD73 / NT5E (Catalog#90192-C08H; Met1-Lys547). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3157