Learn More
Sino Biological Cynomolgus NT5E Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90192C08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus NT5E (EHH53214.1) (Met1-Lys547) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2184