Learn More
Sino Biological Recombinant Cynomolgus TrkA Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 91011C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus NTRK1 (XP_005541454.3) (Met1-Gly423) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-0620