Learn More
Sino Biological Recombinant Anti-LOX-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 90155R014100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus OLR1 / LOX1 (Catalog#90155-C07H; G7PJT2; Ser61-Gln273).
Catalog No. 50-240-8414