Learn More
Sino Biological Anti-LOX-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90155RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus OLR1 / LOX1 (Catalog#90155-C07H; G7PJT2; Ser61-Gln273). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1408