Learn More
Sino Biological Recombinant Cynomolgus OX40 Ligand/TNFSF4 Protein (mFc Tag) 500
Supplier:  Sino Biological 90088C04H500
Encompass_Preferred
A DNA sequence encoding the cynomolgus TNFSF4 (F7FL80) (Gln51-Leu183) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-224-7211