Learn More
Sino Biological Recombinant Cynomolgus PAH Protein (His Tag)
Catalog No. 502992935
Encompass_Preferred
A DNA sequence encoding the Cynomolgus PAH (A0A7N9DEA3) (Asp1-Lys437) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-2935 Supplier Sino Biological Supplier No. 91037C07B1
 Restricted Item
Add to Cart
Add to Cart
A DNA sequence encoding the Cynomolgus PAH (A0A7N9DEA3) (Asp1-Lys437) was expressed, with a polyhistidine tag at the N-terminus.