Learn More
ACROBiosystems Cynomolgus PD-1 25ug
Catalog No. 502534319
Encompass_Preferred
Biotinylated Cynomolgus PD-1 Protein FcAvitag (PD1-C82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).
Catalog No. 50-253-4319 Supplier ACROBiosystems Supplier No. PD1C82F725UG
Biotinylated Cynomolgus PD-1 Protein FcAvitag (PD1-C82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).