Learn More
Sino Biological Recombinant Cynomolgus PD-1 Protein (hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 90311C41HB100
Encompass_Preferred
A DNA sequence encoding the cynomolgus PDCD1 (NP_001271065.1) (Met1-Gln167) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2681