Learn More
Sino Biological Cynomolgus PDGFB Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90030C04H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus PDGFB (EHH65851.1) (Ser82-Thr190) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-237-2724