Learn More
Sino Biological Recombinant Cynomolgus PDGF alpha receptor/PDGFRA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 91008C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus PDGFRA (Q0PW86) (Met1-Glu524) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8820