Learn More
ACROBiosystems Cynomolgus PSMA 200ug
Catalog No. 502534282
Encompass_Preferred
Biotinylated Cynomolgus PSMA Protein HisAvitag (PSA-C82Q6) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # A0A2K5VNZ0).
Catalog No. 50-253-4282 Supplier ACROBiosystems Supplier No. PSAC82Q6200UG
Biotinylated Cynomolgus PSMA Protein HisAvitag (PSA-C82Q6) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # A0A2K5VNZ0).