Learn More
ACROBiosystems PVRIG Protein, Cynomolgus, His Tag, Lyophilized, 100 ug
Catalog No. 502719031
Encompass_Preferred
Cynomolgus PVRIG protein, also known as CD112R, is a transmembrane protein in the poliovirus receptor-like protein family. This protein is expressed from human HEK293 cells and contains the extracellular domain of the target molecule. It features a polyhistidine tag at the C-terminus and is provided in a lyophilized format, reconstituted from a 0.22 μm filtered solution. This protein has been verified for purity and bioactivity, exhibiting binding capability to Nectin-2.
  • Greater than 90% purity by SDS-PAGE and 95% purity by SEC-MALS
  • Endotoxin level less than 0.1 EU per μg
  • Validated binding activity with Nectin-2
  • Calculated molecular weight of 15.6 kDa, migrating at 20-27 kDa due to glycosylation
  • Lyophilized in PBS, pH 7.4 with trehalose
Catalog No. 50-271-9031 Supplier ACROBiosystems Supplier No. PVGC52H3100UG
Cynomolgus PVRIG protein, also known as CD112R, is a transmembrane protein in the poliovirus receptor-like protein family. This protein is expressed from human HEK293 cells and contains the extracellular domain of the target molecule. It features a polyhistidine tag at the C-terminus and is provided in a lyophilized format, reconstituted from a 0.22 μm filtered solution. This protein has been verified for purity and bioactivity, exhibiting binding capability to Nectin-2.
  • Greater than 90% purity by SDS-PAGE and 95% purity by SEC-MALS
  • Endotoxin level less than 0.1 EU per μg
  • Validated binding activity with Nectin-2
  • Calculated molecular weight of 15.6 kDa, migrating at 20-27 kDa due to glycosylation
  • Lyophilized in PBS, pH 7.4 with trehalose