Learn More
ACROBiosystems ACROBiosystems Cynomolgus PVRIG Protein, Mouse IgG2a Fc Tag (MALS verified)
Catalog No. 502106234
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus PVRIG, Mouse IgG2a Fc Tag (PVG-C5253) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # A0A2K5WVV8-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 μg/mL (100 μL/well) can bind Cynomolgus PVRIG, Mouse IgG2a Fc Tag (Cat. No. PVG-C5253) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-210-6234 Supplier ACROBiosystems Supplier No. PVGC52531MG
Recombinant Protein;1MG;Cynomolgus PVRIG, Mouse IgG2a Fc Tag (PVG-C5253) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # A0A2K5WVV8-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 μg/mL (100 μL/well) can bind Cynomolgus PVRIG, Mouse IgG2a Fc Tag (Cat. No. PVG-C5253) with a linear range of 2-20 ng/mL (QC tested).