Learn More
Sino Biological Anti-RBP4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90220T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus RBP4 (Catalog#90220-C08H; Met1-Leu201). RBP4 specific IgG was purified by Cynomolgus RBP4 affinity chromatography.
Catalog No. 50-240-8802