Learn More
Sino Biological Anti-REG1A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90162T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus REG1A (Catalog#90162-C08H; G8F4F8; Met1-Asn166). REG1A specific IgG was purified by Cynomolgus REG1A affinity chromatography.
Catalog No. 50-255-2715