Learn More
Sino Biological Cynomolgus REG1A Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90162C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus REG1A (G8F4F8) (Met1-Asn166) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1748