Learn More
Sino Biological Cynomolgus / Rhesus Angiopoietin-2 / ANG2 Protein (His Tag) 1mg
Supplier:  Sino Biological 90026C07H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus ANGPT2 (XP_005562586.1/XP_001097949.1) (Lys274-Phe495) was expressed with a polyhistidine tag at the N-terminus. Cynomolgus and Rhesus ANGPT2 sequences are identical.
Catalog No. 50-224-7276