Learn More
Sino Biological Recombinant Cynomolgus, Rhesus CD19 Protein (His Tag)
Supplier:  Sino Biological 90051C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD19 (XP_005591597.1) (Met1-Lys292) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD19 sequences are identical.
Catalog No. 50-255-2734