Learn More
Sino Biological Cynomolgus / Rhesus CD19 Protein (His Tag), Biotinylated
Supplier:  Sino Biological 90051C08HB100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD19 ( XP_005591597.1) (Met1-Lys292) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD19 sequences are identical. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6237