Learn More
Sino Biological Recombinant Cynomolgus, Rhesus PD-L1/B7-H1 Protein (hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 90251C41HB100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus / Rhesus CD274 (XP_015292694.1) (Met1-Thr239) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2697