Learn More
Sino Biological Cynomolgus / Rhesus PD-L1 / B7-H1 / CD274 Protein (His Tag), Biotinylated
Supplier:  Sino Biological 90251C08HB100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD274 (XP_015292694.1) (Met1-Thr239) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD274 sequences are identical. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2699