Learn More
Sino Biological Recombinant Cynomolgus, Rhesus PD-L1/B7-H1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 90251C49HB20
Encompass_Preferred
A DNA sequence encoding the Cynomolgus / Rhesus PD-L1 (XP_015292694.1)(Met1-Thr239) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2694