Learn More
Sino Biological Recombinant Cynomolgus, Rhesus Siglec-3/CD33 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 90303C08HB100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD33 (XP_005590138.1) (Met1-Gly248) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD33 sequences are identical. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2683