Learn More
Sino Biological Recombinant Human Cystatin SA Protein (His Tag)
Supplier:  Sino Biological 11567H08H100
Encompass_Preferred
A DNA sequence encoding the human CST2 (NP_001313.1) (Met 1-Ala 141) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6927