Learn More
Sino Biological Cynomolgus / Rhesus CD33 / Siglec-3 Protein (His Tag) 1mg
Supplier:  Sino Biological 90303C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD33 (EHH59833.1 / EHH30318.1) (Met1-Gly248) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD33 sequences are identical.
Catalog No. 50-222-3838