Learn More
Sino Biological Recombinant Cynomolgus, Rhesus CD47 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 90869C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD47 (XP_005548291.1 & NP_001253446.1) (Met1-Asn142) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD47 sequences are identical.
Catalog No. 50-255-2666