Learn More
Sino Biological Cynomolgus / Rhesus c-MET / HGFR Protein (His Tag) 1mg
Supplier:  Sino Biological 90304C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus/rhesus MET (A0A2K5UM36-1 / NP_001162100.2) (Met1-Thr932) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus MET sequences are identical.
Catalog No. 50-224-6982