Learn More
Sino Biological Recombinant Cynomolgus, Rhesus c-MET/HGFR Protein (hFc Tag) 1
Supplier:  Sino Biological 90304C02H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus MET (NP_001162100.1) (Met1-Thr932) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus MET sequences are identical.
Catalog No. 50-224-6666