Learn More
Sino Biological Cynomolgus / Rhesus CD64/FCGR1  Protein (His Tag)
Catalog No. 502993085
Encompass_Preferred
A DNA sequence encoding the Cynomolgus, Rhesus FCGR1 (F7BDL2) (Gln18-Pro288) was expressed with a polyhistidine tag at the C-terminus. The sequences of A0A2K5U901 (Cynomolgus) (Gln33–Pro305) and F7BDL2 (Rhesus macaque) (Gln18–Pro288) are identical.
Catalog No. 50-299-3085 Supplier Sino Biological Supplier No. 91073C08H100
A DNA sequence encoding the Cynomolgus, Rhesus FCGR1 (F7BDL2) (Gln18-Pro288) was expressed with a polyhistidine tag at the C-terminus. The sequences of A0A2K5U901 (Cynomolgus) (Gln33–Pro305) and F7BDL2 (Rhesus macaque) (Gln18–Pro288) are identical.