Learn More
Sino Biological Recombinant Cynomolgus, Rhesus FGFR3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90313C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus FGFR3 (XP_005554344.1) (Met1-Gly375) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus FGFR3 sequences are identical.
Catalog No. 50-228-3312