Learn More
Sino Biological Recombinant Cynomolgus, Rhesus FOLR1 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 90950C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus FOLR1 (A0A2K5UR19 & NP_001181576.1) (Met1-Ser234) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus FOLR1 sequences are identical.
Catalog No. 50-224-6740