Learn More
Sino Biological Recombinant Cynomolgus, Rhesus Fractalkine/CX3CL1 Protein (His Tag) 1
Supplier:  Sino Biological 90316C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CX3CL1 (EHH60415.1) (Met1-Gly100) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CX3CL1 sequences are identical.
Catalog No. 50-224-7233