Learn More
Sino Biological Recombinant Cynomolgus, Rhesus GPRC5D Protein (Nanodisc, His & FLAG Tag)
Catalog No. 502992765
Encompass_Preferred
A DNA sequence encoding the Cynomolgus / Rhesus GPRC5D (A0A2K5W6I7) (Met1-Ser268) was expressed and linked by a Fusion Protein, with a Flag tag at the N-terminus and a polyhistidine tag at the C-terminus. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.
Catalog No. 50-299-2765 Supplier Sino Biological Supplier No. 91022C18HNA100
A DNA sequence encoding the Cynomolgus / Rhesus GPRC5D (A0A2K5W6I7) (Met1-Ser268) was expressed and linked by a Fusion Protein, with a Flag tag at the N-terminus and a polyhistidine tag at the C-terminus. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.