Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque B7-1 / CD80 Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CD0C5252100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque B7-1, Fc Tag (CD0-C5252) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.5 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-1, Fc Tag (Cat. No. CD0-C5252) with a linear range of 1-8 ng/mL (QC tested).
Catalog No. 50-210-6771