Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque B7-1 / CD80 Protein, His Tag (MALS verified)
Catalog No. 502106772
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque B7-1, His Tag (CD0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, Fc Tag (Cat. No. CT4-C5256) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-1, His Tag (Cat. No. CD0-C52H3) with a linear range of 10-156 ng/mL (QC tested).;Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-1, His Tag (Cat. No. CD0-C52H3) with a linear range of 10-156 ng/mL (Routinely tested).
Catalog No. 50-210-6772 Supplier ACROBiosystems Supplier No. CD0C52H3100UG
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque B7-1, His Tag (CD0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, Fc Tag (Cat. No. CT4-C5256) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-1, His Tag (Cat. No. CD0-C52H3) with a linear range of 10-156 ng/mL (QC tested).;Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-1, His Tag (Cat. No. CD0-C52H3) with a linear range of 10-156 ng/mL (Routinely tested).