Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, His Tag
Catalog No. 502106442
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque BCMA, His Tag (BCA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.9 kDa. The protein migrates as 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque BCMA, His Tag (Cat. No. BCA-C52H7) at 2 μg/mL (100 μL/well) can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-210-6442 Supplier ACROBiosystems Supplier No. BCAC52H7100UG
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque BCMA, His Tag (BCA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.9 kDa. The protein migrates as 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque BCMA, His Tag (Cat. No. BCA-C52H7) at 2 μg/mL (100 μL/well) can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.2-3 ng/mL (QC tested).