Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, Fc Tag
Catalog No. 502106439
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque BCMA, Fc Tag (Cat. No. BCA-C5253) with a linear range of 0.05-2 ng/mL (QC tested).;Immobilized Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque BCMA, Fc Tag (Cat. No. BCA-C5253) with a linear range of 0.3-5 ng/mL (Routinely tested).
Catalog No. 50-210-6439 Supplier ACROBiosystems Supplier No. BCAC52531MG
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque BCMA, Fc Tag (Cat. No. BCA-C5253) with a linear range of 0.05-2 ng/mL (QC tested).;Immobilized Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque BCMA, Fc Tag (Cat. No. BCA-C5253) with a linear range of 0.3-5 ng/mL (Routinely tested).