Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque Cadherin-6 / KCAD / CDH6 Protein, His Tag
Catalog No. 502106253
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque Cadherin-6, His Tag (CA6-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Gly 614 (Accession # G7MUQ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.9 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6253 Supplier ACROBiosystems Supplier No. CA6C52H31MG
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque Cadherin-6, His Tag (CA6-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Gly 614 (Accession # G7MUQ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.9 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.