Learn More
ACROBiosystems Cynomolgus / Rhesus macaque CD19 (20-292) Protein, His Tag
Catalog No. 502054921
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque CD19 (20-292), His Tag (CD9-C5221) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 292 (Accession # F7F486-1).; The protein has a calculated MW of 32.0 kDa.
Catalog No. 50-205-4921 Supplier ACROBiosystems Supplier No. CD9C52211MG
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque CD19 (20-292), His Tag (CD9-C5221) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 292 (Accession # F7F486-1).; The protein has a calculated MW of 32.0 kDa.