Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque TIGIT Protein, Mouse IgG2a Fc Tag (MALS verified)
Catalog No. 502106231
Encompass_Preferred
Recombinant Protein;50UG;Cynomolgus / Rhesus macaque TIGIT, Mouse IgG2a Fc Tag (TIT-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # G7NXM4-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD155, Fc,Avitag (Cat. No. CD5-M82F7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Cynomolgus / Rhesus macaque TIGIT, Mouse IgG2a Fc Tag (Cat. No. TIT-C5253) with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 50-210-6231 Supplier ACROBiosystems Supplier No. TITC525350UG
Recombinant Protein;50UG;Cynomolgus / Rhesus macaque TIGIT, Mouse IgG2a Fc Tag (TIT-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # G7NXM4-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD155, Fc,Avitag (Cat. No. CD5-M82F7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Cynomolgus / Rhesus macaque TIGIT, Mouse IgG2a Fc Tag (Cat. No. TIT-C5253) with a linear range of 0.6-20 ng/mL (QC tested).