Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque TPBG / 5T4 Protein, His Tag
Catalog No. 502106897
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque TPBG, His Tag (TPG-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 35 - Ser 355 (Accession # Q4R8Y9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.7 kDa.
Catalog No. 50-210-6897 Supplier ACROBiosystems Supplier No. TPGC52H3100UG
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque TPBG, His Tag (TPG-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 35 - Ser 355 (Accession # Q4R8Y9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.7 kDa.