Learn More
Sino Biological Cynomolgus, Rhesus MET Protein 20ug (Sino Biological)
Supplier:  Sino Biological 90304C08HB20
Encompass_Preferred
A DNA sequence encoding the cynomolgus/rhesus MET (A0A2K5UM36-1 / NP_001162100.2) (Met1-Thr932) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus MET sequences are identical. The purified protein was biotinylated in vitro.
Catalog No. 50-237-2692