Learn More
Sino Biological Recombinant Cynomolgus, Rhesus TL1A/TNFSF15 Protein (His Tag), Trimer, MALS-verified
Catalog No. 502993046
Encompass_Preferred
A DNA sequence encoding the Cynomolgus/Rhesus TNFSF15 (F6S8F9) (Leu72-Leu251) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.
Catalog No. 50-299-3046 Supplier Sino Biological Supplier No. 91015C07H1100
 Restricted Item
Add to Cart
Add to Cart
A DNA sequence encoding the Cynomolgus/Rhesus TNFSF15 (F6S8F9) (Leu72-Leu251) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.