Learn More
Sino Biological Recombinant Cynomolgus, Rhesus B7-H4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90901C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus/rhesus VTCN1 (A0A2K5U6P5-1 & F7B770-1) (Phe29-Ala258) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2659