Learn More
Sino Biological Recombinant Cynomolgus Syndecan-1/CD138 Protein (hFc Tag)
Supplier:  Sino Biological 90938C02H200
Encompass_Preferred
A DNA sequence encoding the cynomolgus SDC1 (XP_005576533.1) (Met1-Glu251) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2644