Learn More
Sino Biological Anti-E-Selectin/CD62E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90169T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus E-Selectin / CD62e / SELE (Catalog#90169-C08H; G8F370; Met1-Pro556). E-Selectin / CD62e / SELE specific IgG was purified by Cynomolgus E-Selectin / CD62e / SELE affinity chromatography.
Catalog No. 50-241-0226