Learn More
Sino Biological Recombinant Cynomolgus E-Selectin/CD62E Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90169C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus SELE (G8F370) (Met1-Pro556) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3444