Learn More
Sino Biological Recombinant Cynomolgus SEZ6 Protein (His Tag), HPLC-verified
Catalog No. 502992763
Encompass_Preferred
A DNA sequence encoding the Cynomolgus SEZ6 (A0A2K5WPJ4-1) (Leu20-His927) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2763 Supplier Sino Biological Supplier No. 91067C08H100
A DNA sequence encoding the Cynomolgus SEZ6 (A0A2K5WPJ4-1) (Leu20-His927) was expressed with a polyhistidine tag at the C-terminus.