Learn More
ACROBiosystems Cynomolgus Siglec-10 Protein, His Tag
Supplier:  ACROBiosystems SI0C52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Siglec-10, His Tag (SI0-C52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - Ala 549 (Accession # A0A2K5WBX8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-205-5270